Iron(II) sulfate heptahydrate

Supplier: USP
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

1270355
101095-002EA 435.38 USD
101095-002
Iron(II) sulfate heptahydrate
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR