Ethylvanillin

Supplier: USP

1267500
101094-984EA 444.62 USD
101094-984
Ethylvanillin
Ethylvanillin
Formula: C₉H₁₀O₃
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 75…77 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR