Ethyl 4-hydroxybenzoate

Supplier: USP

Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate

1267000
101094-982EA 489.23 USD
101094-982
Ethyl 4-hydroxybenzoate
Ethyl 4-hydroxybenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 297
Melting Pt: 116…118 °C
Density: 1.078 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR