Diethyl phthalate

Supplier: USP

1193505
101094-812EA 489.23 USD
101094-812
Diethyl phthalate
Diethyl phthalate
Formula: C₁₂H₁₄O₄
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR