Diethylene glycol monoethyl ether

Supplier: USP

1193301
101094-810EA 469.23 USD
101094-810
Diethylene glycol monoethyl ether
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR