Dibutyl phthalate

Supplier: USP
Danger

Synonyms: DBP, Phthalic acid dibutyl ester

1187080
101094-796EA 453.85 USD
101094-796
Dibutyl phthalate
Dibutyl phthalate
Formula: C₁₆H₂₂O₄
MW: 278.35 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 171 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009441
CAS Number: 84-74-2
EINECS: 201-557-4
UN: 3082
ADR: 9,III
REACH: 01-2119493042-44
Merck Index: 13,01586
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd0889645c3647a1b051c82c9c74ea035c4e295&vid=561e5c60f906bd1eb64b32466d8325825293fda4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR