Dextran, VO Marker

Supplier: USP

1179800
101094-782EA 498.46 USD
101094-782
Dextran, VO Marker
Dextran
Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR