Dexamethasone-21-acetate

Supplier: USP

1176506
101094-754EA 435.38 USD
101094-754
Dexamethasone-21-acetate
Dexamethasone 21-acetate
Formula: C₂₄H₃₁FO₆
MDL Number: MFCD00027407
CAS Number: 1177-87-3
EINECS: 214-646-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR