D-Cycloserine

Supplier: USP
Warning

Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine

1158005
101094-718EA 403.08 USD
101094-718
D-Cycloserine
D-Cycloserine
Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR