Cottonseed oil

Supplier: USP

1150207
101094-686EA 447.69 USD
101094-686
Cottonseed oil
Cottonseed oil
Density: 0,91 g/cm³ (20 °C)
MDL Number: MFCD00130872
CAS Number: 8001-29-4
EINECS: 232-280-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR