Corn oil

Supplier: USP

1148806
101094-682EA 412.31 USD
101094-682
Corn oil
Corn oil
Melting Pt: –10 °C
Density: 0,918 g/cm³ (20 °C)
MDL Number: MFCD00130865
CAS Number: 8001-30-7
EINECS: 232-281-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR