Colchicine

Supplier: USP
Danger

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

1146006
101094-672EA 429.23 USD
101094-672
Colchicine
Colchicine
Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: 154…156 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1 I
Merck Index: 12,02536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR