Clioquinol

Supplier: USP
Danger

Synonyms: 5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol

1138201
101094-648EA 443.08 USD
101094-648
Clioquinol
Clioquinol
Formula: C₉H₅ClINO
MW: 305.5
Boiling Pt: 365
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006787
CAS Number: 130-26-7
EINECS: 204-984-4
UN: 2811
ADR: 6.1 III
Merck Index: 12,05052

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR