Citric acid

Supplier: USP
Warning

1134368
101094-638EA 489.23 USD
101094-638
Citric acid
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1.665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR