Choline chloride

Supplier: USP

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

1133547
101094-618EA 444.62 USD
101094-618
Choline chloride
Choline chloride
Formula: HOH₂CCH₂N(CH₃)₃Cl
MW: 139.62 g/mol
Melting Pt: 244…247 °C
Density: ∼1.205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR