1-Cetylpyridinium chloride monohydrate

Supplier: USP
Danger

1104006
101094-574EA 421.54 USD
101094-574
1-Cetylpyridinium chloride monohydrate
1-Cetylpyridinium chloride monohydrate
Formula: C₂₁H₃₈ClN·1H₂O
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,I
Merck Index: 13,02041

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR