Carbachol

Supplier: USP

1092009
101094-490EA 415.38 USD
101094-490
Carbachol
Carbachol
Formula: H₂NC(O)O(CH₂)₂N(CH₃)₃Cl
Melting Pt: 207 °C
Density: 1.389 g/cm³ (20 °C)
MDL Number: MFCD00012011
CAS Number: 51-83-2
EINECS: 200-127-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01786

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR