Calcium carbonate

Supplier: USP

Synonyms: Marble

1086403
101094-462EA 420 USD
101094-462
Calcium carbonate
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR