Calcium acetate

Supplier: USP

1086334
101094-458EA 435.38 USD
101094-458
Calcium acetate
Calcium acetate
Formula: (H₃CCOO)₂Ca
Density: 1.509 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150019
CAS Number: 62-54-4
EINECS: 200-540-9
Merck Index: 12,01683

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR