Caffeine

Supplier: USP
Warning

Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

1085003
101094-456EA 435.38 USD
101094-456
Caffeine
Caffeine
Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Melting Pt: 235…239 °C
Density: 1.23 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
Merck Index: 13,01636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e609e1c7a199176b9e5b938ef241d166d502f9&vid=1ac1145bba3574e2401439da90f37fee7fd7610b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR