D-(+)-Biotin

Supplier: USP

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

1071508
101094-408EA 444.62 USD
101094-408
D-(+)-Biotin
D(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR