Benzyl benzoate

Supplier: USP
Warning

Synonyms: Benzoic acid benzyl ester

1062008
101094-384EA 435.38 USD
101094-384
Benzyl benzoate
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323
Melting Pt: 21
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9 III
Merck Index: 13,01129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR