L(+)-Arginine

Supplier: USP
Warning

1042500
101094-310EA 513.85 USD
101094-310
L(+)-Arginine
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR