Phenazone

Supplier: USP
Warning

1040005
101094-308EA 403.08 USD
101094-308
Phenazone
Phenazone
Formula: C₁₁H₁₂N₂O
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 152…154 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 12,00757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR