Aluminium hydroxide

Supplier: USP
Warning

1017502
101094-246EA 423.08 USD
101094-246
Aluminium hydroxide
Aluminium hydroxide
Formula: Al(OH)₃
Melting Pt: 300 °C
Density: 2.42 g/cm³ (20 °C)
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7
Merck Index: 12,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR