Ethanol 100%, Reference Standard, USP
Supplier: USP
1012768
101094-230EA 435.38 USD
101094-230
Ethanol 100%, Reference Standard, USP
Standards Organic Standards
  • Total alcohol: ~100%
  • Ethyl alcohol: ~100%
  • Denaturant: none