N,N-Diethyl-m-toluamide

Supplier: USP
Warning

1197007
101094-098EA 435.38 USD
101094-098
N,N-Diethyl-m-toluamide
N,N-Diethyl-m-toluamide
Formula: C₁₂H₁₇NO
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.996 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR