Dexamethasone

Supplier: USP

1176007
101094-090EA 360 USD
101094-090
Dexamethasone
Dexamethasone
Formula: C₂₂H₂₉FO₅
Melting Pt: 268…271 °C
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR