Cellacefate (Cellulose acetate phthalate) USP Reference Standard

Supplier: USP

1098355
101094-062EA 384.62 USD
101094-062
Cellacefate (Cellulose acetate phthalate) USP Reference Standard
Cellacefate (Cellulose acetate phthalate)

Certifications: USP Certified

Formula: C₁₅₆H₁₇₄O₉₅
MDL Number: MFCD00072769
CAS Number: 9004-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR