4-Aminosalicylic acid ≥99%

Supplier: USP
Warning

Synonyms: 4-Amino-2-hydroxybenzoic acid

1026401
101094-038EA 353.85 USD
101094-038
4-Aminosalicylic acid ≥99%
4-Aminosalicylic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 150
Density: 1.545 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007789
CAS Number: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR