Paracetamol

Supplier: USP

1003009
101094-036EA 435.38 USD
101094-036
Paracetamol
Paracetamol
Formula: C₈H₉NO₂
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 168 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 13,00048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR