Anti-SEPSECS Rabbit Polyclonal Antibody

Supplier: Prosci
29-515
10108-446EA 760.88 USD
10108-446
Anti-SEPSECS Rabbit Polyclonal Antibody
Antibodies
SLA/LP converts O-phosphoseryl-tRNA (Sec) to selenocysteinyl-tRNA (Sec) required for selenoprotein biosynthesis.

SLA/LP antibody can be used for detection of SLA/LP by ELISA at 1:312500. SLA/LP antibody can be used for detection of SLA/LP by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SEPSECS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR