Anti-TRNAU1AP Rabbit Polyclonal Antibody

Supplier: Prosci
29-439
10108-230EA 694.55 USD
10108-230
Anti-TRNAU1AP Rabbit Polyclonal Antibody
Antibodies
TRSPAP1 is involved in the early steps of selenocysteine biosynthesis and tRNA (Sec) charging to the later steps resulting in the cotranslational incorporation of selenocysteine into selenoproteins. It stabilizes the SECISBP2, EEFSEC and tRNA (Sec) complex. It may be involved in the methylation of tRNA (Sec) and enhances efficiency of selenoproteins synthesis.

TRSPAP1 antibody can be used for detection of TRSPAP1 by ELISA at 1:312500. TRSPAP1 antibody can be used for detection of TRSPAP1 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRNAU1AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR