CON 2700 Benchtop Meter Package, Oakton® Instruments, Cole Parmer
Supplier: OAKTON INSTRUMENTS MS
WD-35412-00
101076-366EA 1722.2 USD
101076-366
CON 2700 Benchtop Meter Package, Oakton® Instruments, Cole Parmer
Electrochemistry Meters Conductivity Meters
CON 2700 benchtop meter is used for the measurement of conductivity, total dissolved solids (TDS), salinity, resistivity and temperature.
  • Brilliant backlit display with dynamic stability indication
  • Up to 500 point memory with date-time stamp for GLP
  • Output data to printer or PC
  • Accepts either 2-cell or 4-cell electrodes
  • Features auto-standardization, auto-calibration, and autoranging
  • Calibration due alarm with password protection for setup and calibration
  • Measures salinity from 0.0 to 80 ppt and resistitivy from 2.000 ohms to 20.0 Mohms
The CON 2700 meter offers a new compact design that is over 40% smaller than other benchtop meters. The large display is easy to read even at a distance and features stability indication. The meter includes automatic or manual temperature compensation, temperature in °F or °C and display Hold. A calibration due alarm function means no more out-dated calibrations. The water-resistant membrane keypad offers password protection for both set-up and calibration. Multiple pH slopes and offset display enables quick, easy electrode diagnosis. The electrode holder can be mounted on either side of the meter.