Anti-ANP32A Rabbit Polyclonal Antibody

Supplier: Prosci
29-210
10107-774EA 694.55 USD
10107-774
Anti-ANP32A Rabbit Polyclonal Antibody
Antibodies
ANP32A is a novel potent heat-stable inhibitor protein of protein phosphatase 2A. It may play a key role in self-renewing cell populations where it may act in the nucleus to limit their sensitivity to transformation. Being a tumor suppressor, ANP32A represses cell growth through inhibition of transcription by blocking acetylation and phosphorylation of histone H3 and initiating its proapoptotic activity.

ANP32A antibody can be used for detection of ANP32A by ELISA at 1:312500. ANP32A antibody can be used for detection of ANP32A by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANP32A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR