Anti-HSPB1 Rabbit Polyclonal Antibody

Supplier: Prosci
27-301
10104-710EA 694.55 USD
10104-710
Anti-HSPB1 Rabbit Polyclonal Antibody
Antibodies
HSPB1, like the other heat shock proteins, is part of a complex system of molecular chaperones in epidermal keratinocytes.

HSPB1 antibody can be used for detection of HSPB1 by ELISA at 1:62500. HSPB1 antibody can be used for detection of HSPB1 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR