Anti-ZNF766 Rabbit Polyclonal Antibody

Supplier: Prosci
26-518
10103-766EA 760.88 USD
10103-766
Anti-ZNF766 Rabbit Polyclonal Antibody
Antibodies
ZNF766 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 10 C2H2-type zinc fingers and 1 KRAB domain. ZNF766 may be involved in transcriptional regulation.

ZNF766 antibody can be used for detection of ZNF766 by ELISA at 1:1562500. ZNF766 antibody can be used for detection of ZNF766 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF766
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR