Anti-SEP15 Rabbit Polyclonal Antibody

Supplier: Prosci
26-303
10103-426EA 760.88 USD
10103-426
Anti-SEP15 Rabbit Polyclonal Antibody
Antibodies
SEP15 is a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. Studies in mouse suggest that this selenoprotein may have redox function and may be involved in the quality control of protein folding. The gene that encodes the protein is localized on chromosome 1p31, a genetic locus commonly mutated or deleted in human cancers. This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Studies in mouse suggest that this selenoprotein may have redox function and may be involved in the quality control of protein folding. This gene is localized on chromosome 1p31, a genetic locus commonly mutated or deleted in human cancers. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Septin 15 antibody can be used for detection of Septin 15 by ELISA at 1:312500. Septin 15 antibody can be used for detection of Septin 15 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SEP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR