Anti-ATP6V0A2 Rabbit Polyclonal Antibody

Supplier: Prosci
25-762
10102-586EA 760.88 USD
10102-586
Anti-ATP6V0A2 Rabbit Polyclonal Antibody
Antibodies
The multisubunit vacuolar-type proton pump (H (+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H (+)-ATPases are comprised of a peripheral V (1) domain and an integral membrane V (0) domain; ATP6V0A2 is a component of the V (0) domain.The multisubunit vacuolar-type proton pump (H (+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H (+)-ATPases are comprised of a peripheral V (1) domain and an integral membrane V (0) domain; ATP6V0A2 is a component of the V (0) domain (Smith et al., 2003 [PubMed 14580332]).

ATP6V0A2 antibody can be used for detection of ATP6V0A2 by ELISA at 1:312500. ATP6V0A2 antibody can be used for detection of ATP6V0A2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATP6V0A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR