Anti-SRPR Rabbit Polyclonal Antibody

Supplier: Prosci
25-615
10102-358EA 760.88 USD
10102-358
Anti-SRPR Rabbit Polyclonal Antibody
Antibodies
SRPR belongs to the GTP-binding SRP family. It is in conjunction with SRP, the correct targeting of the nascent secretory proteins to the endoplasmic reticulum membrane system.

SRPR antibody can be used for detection of SRPR by ELISA at 1:62500. SRPR antibody can be used for detection of SRPR by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SRPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR