Adapter, Thermocouple with 1:5 PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
5272-15 5274-22 5274-43 5272-17
89051-544EA 219.34 USD
89051-544 89051-546 89051-548 10098-748
Adapter, Thermocouple with 1:5 PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Thermocouple adapter with 1:5 PTFE stopcock with hose connection.
  • Stopcock with hose connection
  • Fabricated from borosilicate glass
Supplied with bushing and FETFE O-ring which allows for compression seal with probes.