Thermocouple Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5041-02 5041-10 5041-114 5041-124
10098-728EA 96.8 USD
10098-728 10098-730 10098-732 10098-734
Thermocouple Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This thermocouple adapter has a lower extension to better help support the thermocouple.
  • With lower extension
  • Compression-style Ace-Thred nylon bushing
  • PTFE ferrule
Designed for use when thermocouples are bending or appearing to move excesivelly within a joint. The thermocouple is secured with a compression style Ace-Thred nylon bushing and PTFE ferrule.