PermaRed/AP Auto, Diagnostic BioSystems
Supplier: Diagnostic Biosystems
K049-AUTO-110
10098-720EA 545.09 USD
10098-720
PermaRed/AP Auto, Diagnostic BioSystems
Assays Cellular Assays
PermaRed/AP-Auto produces a red color, ranging from pink to brilliant dark red, depending on the signal intensity
PermaRed/AP-Auto has been formulated for on-slide mixing. The instrument should be programmed to apply the substrate-buffer to the slide followed by application of the chromogen reagent. The substrate-buffer and chromogen components are stable in their separate formats. They can be loaded into the reagent rack at the beginning of the staining run with the other reagents, for walk away convenience.
It is insoluble in organic solvents; therefore sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted.