CC/Mount™ Aqueous Permanent Mounting Medium, Diagnostic Biosystems
Supplier: Diagnostic Biosystems.jpg)
CC/mount is an aqueous mounting medium with a high refractive index. When applied to stained tissue sections, specimens can be permanently mounted without chromogens fading.
The advantages of CC/mount include no coverslip, no exposure to organic fumes, permanent storage of slides and high resolution of tissue specimens. CC/Mount is compatible with chromogens like AEC, DAB, Fast Red, BCIP/NBT, BCIP/INT and fluorescent dyes like FITC and phycobiliproteins. The high pH ensures increased stability of fluorescence.