Anti-PSPN Rabbit Polyclonal Antibody

Supplier: Prosci
38-119
10098-228EA 587.65 USD
10098-228
Anti-PSPN Rabbit Polyclonal Antibody
Antibodies
Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken and was later shown to be the preferential binding subunit for Persephin. In contrast to GDNF and Neurturin, Persephin does not support survival of peripheral neurons.

ELISA:
Indirect:
To detect hPersephin by indirect ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hPersephin.

Sandwich
To detect hPersephin by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.5 - 2.0 ?g/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with our biotinylated Anti-Human Persephin as a detection antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hPersephin.

Western Blot:
To detect hPersephin by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hPersephin is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.

The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.

Type: Primary
Antigen: PSPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR