Anti-ZNF671 Rabbit Polyclonal Antibody

Supplier: Proteintech
21329-1-AP-150UL
10097-370EA 462.47 USD
10097-370
Anti-ZNF671 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: ZNF671
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR