Anti-ZNF391 Rabbit Polyclonal Antibody

Supplier: Proteintech
24805-1-AP-150UL
10097-346EA 462.47 USD
10097-346
Anti-ZNF391 Rabbit Polyclonal Antibody
Antibodies
ZNF391 is involved in transcriptional regulation. The calculated MW of ZFN391 is 358aa, 41 kDa. We always got 60-70 kDa band in our detection. The immunogen of this antibody is 1-128aa of human ZNF391. No other protein can be recognized by this antibody from the sequence BLAST results.

IP: Mouse Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: ZNF391
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR