Anti-ZMPSTE24 Rabbit Polyclonal Antibody

Supplier: Proteintech
12847-1-AP-150UL
10097-294EA 462.47 USD
10097-294
Anti-ZMPSTE24 Rabbit Polyclonal Antibody
Antibodies
ZMPSTE24(CAAX prenyl protease 1 homolog) gene encodes a zinc metalloproteinase involved in the processing of farnesylated proteins(PubMed: 10373325). The significance of ZMPSTE24 in human disease stems from its role as an enzyme necessary for the correct processing and maturation of lamin A ,an intermediate filament component of the nuclear envelope (PubMed: 16297189).

  • Western Blot:PC-3 Cells, 1:200-1:2000; IF:PC-3 Cells, 1:10-1:100

Type: Primary
Antigen: ZMPSTE24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR