Anti-ZFYVE16 Rabbit Polyclonal Antibody

Supplier: Proteintech
13118-2-AP-150UL
10097-272EA 462.47 USD
10097-272
Anti-ZFYVE16 Rabbit Polyclonal Antibody
Antibodies
Endofin (also known as ZFYVE16) is an endosomal FYVE domian protein implicated in regulating membrane trafficking. Endofin has been shown to localize to early endosomes. It co-localizes with SARA but does not associate with it or Smad2 and does not behave like SARA in affecting TGF-signaling. Northern blot analysis showed ZFYVE16 was widely expressed with high levels in kidney, placenta and lung. Overexpression of Endofin causes endosome aggregation. The gene of ZFYVE16 maps to chromosome 5q14, and encodes a 1539-amino acid protein with a molecular mass of 169 kDa. In addition, endogenous Endofin can also be detected as a band of 230-250 kDa.

  • Western Blot: MDA-MB-453s Cells, 1:500-1:5000; IP: HeLa Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: ZFYVE16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR