Anti-ZCCHC6 Rabbit Polyclonal Antibody

Supplier: Proteintech
25196-1-AP-150UL
10097-260EA 462.47 USD
10097-260
Anti-ZCCHC6 Rabbit Polyclonal Antibody
Antibodies
ZCCHC6 also named as KIAA1711 ,HS2 or TUT7 is a 1495 amino acid protein, which contains threes CCHC-type zinc fingers and two PAP-associated domains. A member of the DNA polymerase type-B-like family, ZCCHC6 as a uridylyltransferase that mediates RNA uridylation.

Western Blot:PC-3 Cells, 1:500-1:5000; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: ZCCHC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR